Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285742.1 | NP_001272671.1 | 4 - 5 | 635 | 62 | |
| GenBank mRNA | GU647086.1 | - | 4 - 5 | 635 | 62 | |
| JO592165.2 | - | 3 - 4 | 589 | 62 | ||
More Information
Set Membership: |
|

NCR1
Genomic Map