Assay Details
| Gene Symbol: KRTAP3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285774.1 | NP_001272703.1 | 1 | 226 | 84 | |
| GenBank mRNA | AY510120.1 | - | 1 | 227 | 84 | |
| KA306222.1 | - | 1 | 317 | 84 | ||
More Information
Set Membership: |
|

KRTAP3-1
Genomic Map