Assay Details
| Gene Symbol: OAZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290120.1 | NP_001277049.1 | 4 - 5 | 648 | 76 | |
| GenBank mRNA | JO318199.2 | - | 4 - 5 | 603 | 76 | |
| JO631734.2 | - | 4 - 5 | 648 | 76 | ||
| JR119748.1 | - | 4443 | 76 | |||
| KA314417.1 | - | 4 - 5 | 747 | 76 | ||
More Information
Set Membership: |
|

OAZ2
Genomic Map