Assay Details
| Gene Symbol: CSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285570.1 | NP_001272499.1 | 1 - 2 | 153 | 87 | |
| XM_013965265.2 | XP_013820719.1 | 1 - 2 | 176 | 87 | ||
| GenBank mRNA | DQ010419.1 | - | 1 - 2 | 153 | 87 | |
More Information
Set Membership: |
|

CSF2
Genomic Map