Assay Details
| Gene Symbol: CHI3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285689.1 | NP_001272618.1 | 7 - 8 | 730 | 66 | |
| XM_005690369.3 | XP_005690426.2 | 7 - 8 | 856 | 66 | ||
| GenBank mRNA | AY081150.1 | - | 7 - 8 | 730 | 66 | |
| JO340909.2 | - | 2 - 3 | 2096 | 66 | ||
More Information
Set Membership: |
|

CHI3L1
Genomic Map