Assay Details
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314268.1 | NP_001301197.1 | 9 - 10 | 960 | 67 | |
| XM_018058064.1 | XP_017913553.1 | 9 - 10 | 1708 | 67 | ||
| GenBank mRNA | JO581586.1 | - | 2 - 3 | 878 | 67 | |
| JQ308780.1 | - | 9 - 10 | 960 | 67 | ||
| JQ308789.1 | - | 9 - 10 | 960 | 67 | ||
More Information
Set Membership: |
|

TRAM1
Genomic Map