Assay Details
| Gene Symbol: FFAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285653.1 | NP_001272582.1 | 1 | 820 | 95 | |
| XM_005692314.3 | XP_005692371.2 | 2 | 1071 | 95 | ||
| GenBank mRNA | HM013824.1 | - | 1 | 820 | 95 | |
More Information
Set Membership: |
|

FFAR3
Genomic Map