Assay Details
| Gene Symbol: RAP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287233.1 | NP_001274162.1 | 7 - 8 | 757 | 92 | |
| XM_018047553.1 | XP_017903042.1 | 7 - 8 | 92 | |||
| GenBank mRNA | JO581186.1 | - | 92 | |||
| JQ417404.1 | - | 7 - 8 | 757 | 92 | ||
More Information
Set Membership: |
|

RAP1B
Genomic Map