Assay Details
| Gene Symbol: MC4R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285591.1 | NP_001272520.1 | 1 | 927 | 74 | |
| GenBank mRNA | JN107563.1 | - | 1 | 647 | 74 | |
| JN580891.1 | - | 1 | 927 | 74 | ||
More Information
Set Membership: |
|

MC4R
Genomic Map