Assay Details
| Gene Symbol: DCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285555.1 | NP_001272484.1 | 2 - 3 | 327 | 65 | |
| XM_005679813.3 | XP_005679870.1 | 2 - 3 | 65 | |||
| GenBank mRNA | HQ326237.1 | - | 2 - 3 | 327 | 65 | |
More Information
Set Membership: |
|

DCN
Genomic Map