Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288818.1 | NP_001275747.1 | 11 - 12 | 1408 | 69 | |
| GenBank mRNA | JT717286.1 | - | 11 - 12 | 1430 | 69 | |
| KF726108.1 | - | 11 - 12 | 1408 | 69 | ||
More Information
Set Membership: |
|

ETV5
Genomic Map