Assay Details
| Gene Symbol: RPLP0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314331.1 | NP_001301260.1 | 3 - 4 | 315 | 88 | |
| XM_005709526.3 | XP_005709583.3 | 4 - 5 | 406 | 88 | ||
| GenBank mRNA | JO581861.1 | - | 4 - 5 | 592 | 88 | |
| KM076930.1 | - | 3 - 4 | 315 | 88 | ||
More Information
Set Membership: |
|

RPLP0
Genomic Map