Assay Details
| Gene Symbol: CXCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681749.3 | XP_005681806.1 | 2 - 3 | 276 | 104 | |
| GenBank mRNA | JT638447.1 | - | 2 - 3 | 257 | 104 | |
More Information
Set Membership: |
|

CXCL8
Genomic Map