Assay Details
| Gene Symbol: STAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013975437.2 | XP_013830891.2 | 4 - 5 | 754 | 69 | |
| GenBank mRNA | KA353390.1 | - | 3 - 4 | 990 | 69 | |
More Information
Set Membership: |
|

STAR
Genomic Map