Assay Details
| Gene Symbol: CFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043098.1 | XP_017898587.1 | 2 - 3 | 544 | 75 | |
| GenBank mRNA | JO580982.1 | - | 2 - 3 | 718 | 75 | |
| KA327339.1 | - | 2 - 3 | 468 | 75 | ||
More Information
Set Membership: |
|

CFL1
Genomic Map