Assay Details
| Gene Symbol: IL1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686665.3 | XP_005686722.2 | 4 - 5 | 413 | 70 | |
| GenBank mRNA | D63350.1 | - | 3 - 4 | 334 | 70 | |
More Information
Set Membership: |
|

IL1A
Genomic Map