Assay Details
| Gene Symbol: ZDHHC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691844.3 | XP_005691901.1 | 7 - 8 | 981 | 67 | |
| XM_018061802.1 | XP_017917291.1 | 7 - 8 | 1081 | 67 | ||
| XM_018061803.1 | XP_017917292.1 | 7 - 8 | 860 | 67 | ||
| GenBank mRNA | JO427124.1 | - | 1 - 2 | 1888 | 67 | |
| KA346966.1 | - | 1 - 2 | 1830 | 67 | ||
More Information
Set Membership: |
|

ZDHHC7
Genomic Map