Assay Details
| Gene Symbol: ZNF106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054397.1 | XP_017909886.1 | 16 - 17 | 5375 | 69 | |
| XM_018054398.1 | XP_017909887.1 | 14 - 15 | 2996 | 69 | ||
More Information
Set Membership: |
|

ZNF106
Genomic Map