Assay Details
| Gene Symbol: NDUFAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686495.2 | XP_005686552.2 | 7 - 8 | 844 | 64 | |
| XM_005686496.2 | XP_005686553.1 | 7 - 8 | 851 | 64 | ||
| GenBank mRNA | KA330107.1 | - | 7 - 8 | 837 | 64 | |
More Information
Set Membership: |
|

NDUFAF7
Genomic Map