Assay Details
| Gene Symbol: PCYOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055468.1 | XP_017910957.1 | 2 - 3 | 399 | 60 | |
| GenBank mRNA | JO380133.2 | - | 4 - 5 | 1886 | 60 | |
| KA325722.1 | - | 4 - 5 | 2084 | 60 | ||
More Information
Set Membership: |
|

PCYOX1
Genomic Map