Assay Details
| Gene Symbol: CMPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055703.1 | XP_017911192.1 | 4 - 5 | 60 | ||
| GenBank mRNA | JR122339.1 | - | 2189 | 226 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CMPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055703.1 | XP_017911192.1 | 4 - 5 | 60 | ||
| GenBank mRNA | JR122339.1 | - | 2189 | 226 | ||
Set Membership: |
|