Assay Details
| Gene Symbol: TMEM235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063685.1 | XP_017919174.1 | 3 - 4 | 497 | 67 | |
| GenBank mRNA | KA320855.1 | - | 3 - 4 | 369 | 67 | |
More Information
Set Membership: |
|

TMEM235
Genomic Map