Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048713.1 | XP_017904202.1 | 3 - 4 | 372 | 81 | |
XM_018048714.1 | XP_017904203.1 | 3 - 4 | 457 | 81 | ||
GenBank mRNA | JO396301.2 | - | 1 - 2 | 176 | 81 | |
JO500317.2 | - | 272 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
