Assay Details
| Gene Symbol: ENOPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681827.3 | XP_005681884.1 | 3 - 4 | 665 | 62 | |
| GenBank mRNA | JT663457.1 | - | 7 - 8 | 1628 | 62 | |
| JT713150.1 | - | 5 - 6 | 923 | 62 | ||
More Information
Set Membership: |
|

ENOPH1
Genomic Map