Assay Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687190.3 | XP_005687247.2 | 2 - 3 | 212 | 62 | |
| XM_018055871.1 | XP_017911360.1 | 2 - 3 | 411 | 62 | ||
| XM_018055872.1 | XP_017911361.1 | 2 - 3 | 401 | 62 | ||
| XM_018055873.1 | XP_017911362.1 | 2 - 3 | 290 | 62 | ||
| XM_018055874.1 | XP_017911363.1 | 2 - 3 | 294 | 62 | ||
| XM_018055875.1 | XP_017911364.1 | 2 - 3 | 282 | 62 | ||
| GenBank mRNA | JT680751.1 | - | 2 - 3 | 185 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005687190.3 transcripts.
Set Membership: |
|

COQ4
Genomic Map