Assay Details
| Gene Symbol: LOC102171800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688326.3 | XP_005688383.2 | 2 - 3 | 474 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102171800| Gene Symbol: LOC102171800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688326.3 | XP_005688383.2 | 2 - 3 | 474 | 102 | |
Set Membership: |
|