Assay Details
| Gene Symbol: COLEC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689132.3 | XP_005689189.1 | 4 - 5 | 513 | 60 | |
| GenBank mRNA | JO422188.2 | - | 4 - 5 | 2169 | 60 | |
More Information
Set Membership: |
|

COLEC10
Genomic Map