Assay Details
| Gene Symbol: RIBC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013976546.2 | XP_013832000.1 | 3 - 4 | 3589 | 74 | |
| XM_013976550.2 | XP_013832004.1 | 3 - 4 | 3589 | 74 | ||
| XM_018043904.1 | XP_017899393.1 | 3 - 4 | 3589 | 74 | ||
| XM_018043905.1 | XP_017899394.1 | 3 - 4 | 3589 | 74 | ||
| XM_018043906.1 | XP_017899395.1 | 5 - 6 | 848 | 74 | ||
More Information
Set Membership: |
|

RIBC1
Genomic Map