Assay Details
| Gene Symbol: MED26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051349.1 | XP_017906838.1 | 2 - 3 | 427 | 69 | |
| GenBank mRNA | JO593043.2 | - | 1 - 2 | 2734 | 69 | |
| KA314139.1 | - | 1 - 2 | 2327 | 69 | ||
More Information
Set Membership: |
|

MED26
Genomic Map