Assay Details
| Gene Symbol: HADHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686934.3 | XP_005686991.1 | 2 - 3 | 208 | 67 | |
| XM_013967822.2 | XP_013823276.1 | 2 - 3 | 208 | 67 | ||
| GenBank mRNA | JO375318.2 | - | 14 - 15 | 2014 | 67 | |
| JO428064.1 | - | 14 - 15 | 2361 | 67 | ||
More Information
Set Membership: |
|

HADHB
Genomic Map