Assay Details
| Gene Symbol: MED27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687280.1 | XP_005687337.1 | 2 - 3 | 387 | 59 | |
| GenBank mRNA | JO586886.2 | - | 2 - 3 | 367 | 59 | |
| KA337415.1 | - | 2 - 3 | 373 | 59 | ||
More Information
Set Membership: |
|

MED27
Genomic Map