Assay Details
| Gene Symbol: ACTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005698192.3 | XP_005698249.1 | 7 - 8 | 1298 | 71 | |
| GenBank mRNA | JO503311.2 | - | 2 - 3 | 250 | 71 | |
More Information
Set Membership: |
|

ACTA2
Genomic Map