Assay Details
| Gene Symbol: GNA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050801.1 | XP_017906290.1 | 4 - 5 | 986 | 87 | |
| XM_018050802.1 | XP_017906291.1 | 4 - 5 | 983 | 87 | ||
| GenBank mRNA | JO598101.2 | - | 1 - 2 | 58 | 87 | |
More Information
Set Membership: |
|

GNA15
Genomic Map