Assay Details
Gene Symbol: YIPF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013967524.2 | XP_013822978.1 | 5 - 6 | 850 | 85 | |
GenBank mRNA | JO369253.2 | - | 5 - 6 | 634 | 85 | |
JO435973.2 | - | 5 - 6 | 849 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
