Assay Details
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056256.1 | XP_017911745.1 | 6 - 7 | 1505 | 60 | |
XM_018056258.1 | XP_017911747.1 | 6 - 7 | 1505 | 60 | ||
GenBank mRNA | JO347930.2 | - | 6 - 7 | 1316 | 60 | |
JO388360.2 | - | 7 - 8 | 1154 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
