Assay Details
| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057204.1 | XP_017912693.1 | 6 - 7 | 1379 | 67 | |
| XM_018057205.1 | XP_017912694.1 | 6 - 7 | 1373 | 67 | ||
More Information
Set Membership: |
|

MAP3K8
Genomic Map