Assay Details
| Gene Symbol: DCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681738.3 | XP_005681795.1 | 4 - 5 | 860 | 93 | |
| XM_018049557.1 | XP_017905046.1 | 4 - 5 | 778 | 93 | ||
| GenBank mRNA | KA341652.1 | - | 4 - 5 | 779 | 93 | |
More Information
Set Membership: |
|

DCK
Genomic Map