Assay Details
| Gene Symbol: LOC102183680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682222.3 | XP_005682279.2 | 2 - 3 | 280 | 83 | |
| XM_013965041.2 | XP_013820495.2 | 2 - 3 | 424 | 83 | ||
| XM_013965042.2 | XP_013820496.2 | 2 - 3 | 215 | 83 | ||
| XM_018051320.1 | XP_017906809.1 | 2 - 3 | 428 | 83 | ||
| XM_018051321.1 | XP_017906810.1 | 2 - 3 | 398 | 83 | ||
More Information
Set Membership: |
|

LOC102183680
Genomic Map