Assay Details
| Gene Symbol: LOC102183680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682222.3 | XP_005682279.2 | 3 - 4 | 432 | 74 | |
| XM_013965041.2 | XP_013820495.2 | 3 - 4 | 576 | 74 | ||
| XM_013965042.2 | XP_013820496.2 | 3 - 4 | 367 | 74 | ||
| XM_018051320.1 | XP_017906809.1 | 3 - 4 | 580 | 74 | ||
| XM_018051321.1 | XP_017906810.1 | 3 - 4 | 550 | 74 | ||
More Information
Set Membership: |
|

LOC102183680
Genomic Map