Assay Details
| Gene Symbol: NAPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062859.1 | XP_017918348.1 | 3 - 4 | 774 | 69 | |
| GenBank mRNA | JO599577.2 | - | 3 - 4 | 355 | 69 | |
More Information
Set Membership: |
|

NAPSA
Genomic Map