Assay Details
| Gene Symbol: PIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005700651.2 | XP_005700708.1 | 3 - 4 | 337 | 99 | |
| GenBank mRNA | KA323845.1 | - | 1 - 2 | 171 | 99 | |
More Information
Set Membership: |
|

PIN4
Genomic Map