Assay Details
| Gene Symbol: ZNF276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061906.1 | XP_017917395.1 | 7 - 8 | 1389 | 65 | |
| XM_018061907.1 | XP_017917396.1 | 6 - 7 | 1204 | 65 | ||
| XM_018061908.1 | XP_017917397.1 | 7 - 8 | 1176 | 65 | ||
| XM_018061909.1 | XP_017917398.1 | 7 - 8 | 1163 | 65 | ||
More Information
Set Membership: |
|

ZNF276
Genomic Map