Assay Details
| Gene Symbol: LOC102170758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680264.3 | XP_005680321.2 | 1 - 2 | 533 | 103 | |
| GenBank mRNA | JO444681.2 | - | 1 - 2 | 306 | 103 | |
More Information
Set Membership: |
|

LOC102170758
Genomic Map