Assay Details
| Gene Symbol: LOC102170758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680264.3 | XP_005680321.2 | 4 - 5 | 1132 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102170758| Gene Symbol: LOC102170758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680264.3 | XP_005680321.2 | 4 - 5 | 1132 | 82 | |
Set Membership: |
|