Assay Details
| Gene Symbol: LOC102170758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680264.3 | XP_005680321.2 | 8 - 9 | 1751 | 99 | |
| GenBank mRNA | JT666742.1 | - | 9 - 10 | 1681 | 99 | |
More Information
Set Membership: |
|

LOC102170758
Genomic Map