Assay Details
| Gene Symbol: LOC102172077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688327.3 | XP_005688384.2 | 4 - 5 | 1006 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102172077| Gene Symbol: LOC102172077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688327.3 | XP_005688384.2 | 4 - 5 | 1006 | 71 | |
Set Membership: |
|