Assay Details
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691413.3 | XP_005691470.1 | 5 - 6 | 786 | 54 | |
XM_013970679.2 | XP_013826133.1 | 4 - 5 | 1007 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
