Assay Details
| Gene Symbol: LOC102175889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693218.3 | XP_005693275.1 | 1 - 2 | 426 | 93 | |
| GenBank mRNA | JO368962.2 | - | 1 - 2 | 164 | 93 | |
| JO514448.2 | - | 1 - 2 | 157 | 93 | ||
More Information
Set Membership: |
|

LOC102175889
Genomic Map