Assay Details
| Gene Symbol: METTL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693970.2 | XP_005694027.2 | 7 - 8 | 964 | 68 | |
| GenBank mRNA | KA334515.1 | - | 2 - 3 | 335 | 68 | |
More Information
Set Membership: |
|

METTL2A
Genomic Map