Assay Details
| Gene Symbol: FKBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040070.1 | XP_017895559.1 | 7 - 8 | 958 | 79 | |
| XM_018040071.1 | XP_017895560.1 | 7 - 8 | 900 | 79 | ||
| XM_018040072.1 | XP_017895561.1 | 6 - 7 | 809 | 79 | ||
More Information
Set Membership: |
|

FKBP6
Genomic Map